accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 613-92-3

IUPAC Name: (Z)-N'-hydroxybenzenecarboximidamide
Synonyms: (Z)-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: MXOQNVMDKHLYCZ-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=CC=C1
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)