accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 613-93-4

CH3HNO

CAS RN 613-93-4

IUPAC Name: N-methylbenzamide
Synonyms: N-methylbenzamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: NCCHARWOCKOHIH-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results