CAS RN 6132-17-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fensulfothion Oxon Sulfone, TRC
CAS: 6132-17-8 Molecular Formula: C11 H17 O6 P S Molecular Weight (g/mol): 308.29 Synonym: Phosphoric acid, diethyl 4-(methylsulfonyl)phenyl ester,Phosphoric acid, diethyl p-(methylsulfonyl)phenyl ester (6CI,7CI,8CI),Phenol, p-(methylsulfonyl)-, diethyl phosphate (8CI),Diethyl 4-methylsulfonylphenyl phosphate,Fensulfothion oxon sulfone,Fensulfothion oxygen analog sulfone,O,O-Diethyl O-(4-methylsulfonylphenyl) phosphate IUPAC Name: diethyl (4-methylsulfonylphenyl) phosphate SMILES: CCOP(=O)(OCC)Oc1ccc(cc1)S(=O)(=O)C