accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61341-26-2

CH3SClO

CAS RN 61341-26-2

IUPAC Name: 3-methylthiophene-2-carbonyl chloride
Synonyms: 3-methylthiophene-2-carbonyl chloride
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: AJSVPEVDFBYRCH-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)C(Cl)=O
Boiling Point 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)