accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 613657-61-7

SOOHNOOHFFF

CAS RN 613657-61-7

IUPAC Name: 4-[3-(trifluoromethyl)benzenesulfonamido]benzoic acid
Synonyms: 4-[3-(trifluoromethyl)benzenesulfonamido]benzoic acid
Molecular Weight (g/mol): 345.29
Molecular Formula: C14H10F3NO4S
InChi Key: NJEQBXPASRIDHG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results