accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(R)(R)(R)(R)(S)(S)OOO(S)(S)(R)(R)HOOHOHHOHOOHHOOHH2OH2O

CAS RN 6138-23-4

IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
Synonyms: α,α'-trehalose dihydrate
Molecular Weight (g/mol): 378.33
Molecular Formula: C12H26O13
InChi Key: DPVHGFAJLZWDOC-PVXXTIHASA-N
SMILES: O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (2)
  • (2)
  • (6)
  • (1)
  • (6)
  • (5)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (14)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (4)
  • (6)
  • (1)
Physical Form 
  • (8)
  • (2)
  • (4)
Grade 
  • (2)
  • (6)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (10)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (6)
  • (1)
  • (6)
  • (5)
  • (2)
  • (1)
  • (3)
  • (4)
  • (14)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (6)
  • (1)
  • (8)
  • (2)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)