accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O(E)

CAS RN 614-47-1

IUPAC Name: (2E)-1,3-diphenylprop-2-en-1-one
Synonyms: trans-chalcone
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: DQFBYFPFKXHELB-VAWYXSNFSA-N
SMILES: O=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results