accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHBr

CAS RN 614-76-6

IUPAC Name: N-(2-bromophenyl)acetamide
Synonyms: 2-bromoacetanilide
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: VOBKUOHHOWQHFZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results