accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 614-80-2

IUPAC Name: N-(2-hydroxyphenyl)acetamide
Synonyms: O-hydroxyacetanilide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ADVGKWPZRIDURE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results