accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61425-27-2

CH3(R)OHNHOO

CAS RN 61425-27-2

IUPAC Name: benzyl N-(1-hydroxypropan-2-yl)carbamate
Synonyms: benzyl N-(1-hydroxypropan-2-yl)carbamate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: AFPHMHSLDRPUSM-UHFFFAOYNA-N
SMILES: CC(CO)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)