accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61436-88-2

CH3HNOBr

CAS RN 61436-88-2

IUPAC Name: 2-bromo-N-methylbenzamide
Synonyms: 2-bromo-N-methylbenzamide
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: FKVCJQNPXXXZIE-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results