accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6146-52-7

IUPAC Name: 5-nitro-1H-indole
Synonyms: indole, 5-nitro
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: OZFPSOBLQZPIAV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)