accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OCoCH3OOCH3OO

CAS RN 6147-53-1

IUPAC Name: λ²-cobalt(2+) diacetate tetrahydrate
Synonyms: λ²-cobalt(2+) diacetate tetrahydrate
Molecular Weight (g/mol): 249.08
Molecular Formula: C4H14CoO8
InChi Key: ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES: O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (15)
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (5)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
Physical Form 
  • (7)
  • (3)
  • (2)
  • (1)
Grade 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (13)
  • (7)
  • (15)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (5)
  • (4)
  • (2)
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)