accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 615-36-1

IUPAC Name: 2-bromoaniline
Synonyms: O-bromoaniline
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: AOPBDRUWRLBSDB-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)