accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 615-37-2

IUPAC Name: 1-iodo-2-methylbenzene
Synonyms: O-iodotoluene
Molecular Weight (g/mol): 218.04
Molecular Formula: C7H7I
InChi Key: RINOYHWVBUKAQE-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)