accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

II

CAS RN 615-42-9

IUPAC Name: 1,2-diiodobenzene
Synonyms: 1,2-diiodobenzene
Molecular Weight (g/mol): 329.91
Molecular Formula: C6H4I2
InChi Key: BBOLNFYSRZVALD-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1I
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results