accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 615-57-6

IUPAC Name: 2,4-dibromoaniline
Synonyms: 2,4-dibromoaniline
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: DYSRXWYRUJCNFI-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results