accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 615-74-7

IUPAC Name: 2-chloro-5-methylphenol
Synonyms: 6-chloro-m-cresol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: SMFHPCZZAAMJJO-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(O)=C1
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)