accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOHO

CAS RN 616-30-8

IUPAC Name: 3-aminopropane-1,2-diol
Synonyms: 3-aminopropane-1,2-diol
Molecular Weight (g/mol): 91.11
Molecular Formula: C3H9NO2
InChi Key: KQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILES: NCC(O)CO
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)