accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOH3C

CAS RN 616-38-6

IUPAC Name: dimethyl carbonate
Synonyms: dimethyl carbonate methyl carbonate, 11C-labeled methyl carbonate ((meo)2CO) 11c-labeled methyl carbonate 11C-labeled methyl carbonate methyl carbonate, 11c-labeled
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: IEJIGPNLZYLLBP-UHFFFAOYSA-N
SMILES: COC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (6)
Percent Purity 
  • (7)
  • (2)
Physical Form 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (6)
  • (7)
  • (2)
  • (2)
  • (7)
  • (2)