accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3CH3H3C

CAS RN 6163-64-0

IUPAC Name: 2-methyl-2-(methylsulfanyl)propane
Synonyms: 2-methyl-2-(methylsulfanyl)propane
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: CJFVCTVYZFTORU-UHFFFAOYSA-N
SMILES: CSC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results