accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6164-79-0

IUPAC Name: methyl pyrazine-2-carboxylate
Synonyms: 2-methylpyrazine carboxylate
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: TWIIRMSFZNYMQE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results