accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OBOH3CH3CH3CCH3O

CAS RN 61676-62-8

IUPAC Name: 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane
Synonyms: 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 186.06
Molecular Formula: C9H19BO3
InChi Key: MRWWWZLJWNIEEJ-UHFFFAOYSA-N
SMILES: CC(C)OB1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)