CAS RN 61696-54-6
CAS RN 61696-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic Acid, TRC
CAS: 61696-54-6 Molecular Formula: C16 H24 O14 Molecular Weight (g/mol): 440.35 IUPAC Name: (2R,3R,11R,12R)-1,4,7,10,13,16-hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic acid SMILES: OC(=O)[C@@H]1OCCOCCO[C@H]([C@@H](OCCOCCO[C@H]1C(=O)O)C(=O)O)C(=O)O