accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO

CAS RN 617-02-7

IUPAC Name: 2-methylphenyl benzoate
Synonyms: 2-methylphenyl benzoate
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: YPMKPCAILYDVBN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results