accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH3CN

CAS RN 617-83-4

IUPAC Name: cyanodiethylamine
Synonyms: diethylcyanamide
Molecular Weight (g/mol): 98.15
Molecular Formula: C5H10N2
InChi Key: ZZTSQZQUWBFTAT-UHFFFAOYSA-N
SMILES: CCN(CC)C#N
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results