accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiH3CH3C

CAS RN 617-86-7

IUPAC Name: triethylsilyl
Synonyms: triethylsilane
Molecular Weight (g/mol): 115.27
Molecular Formula: C6H15Si
InChi Key: QXTIBZLKQPJVII-UHFFFAOYSA-N
SMILES: CC[Si](CC)CC
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (4)