accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61713-23-3

IUPAC Name: ethyl 2-({[(2-ethoxy-2-oxoethyl)sulfanyl]methyl}sulfanyl)acetate
Synonyms: ethyl 2-({[(2-ethoxy-2-oxoethyl)sulfanyl]methyl}sulfanyl)acetate
Molecular Weight (g/mol): 252.34
Molecular Formula: C9H16O4S2
InChi Key: NBNGVIRHOBZBHM-UHFFFAOYSA-N
SMILES: CCOC(=O)CSCSCC(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results