accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6175-49-1

H3CCH3O

CAS RN 6175-49-1

IUPAC Name: dodecan-2-one
Synonyms: methyl decyl ketone
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: LSKONYYRONEBKA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)