accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 617718-46-4

IUPAC Name: 3-fluoroazetidine hydrochloride
Synonyms: 3-fluoroazetidine hydrochloride
Molecular Weight (g/mol): 111.54
Molecular Formula: C3H7ClFN
InChi Key: PXFUWRWCKSLCLS-UHFFFAOYSA-N
SMILES: Cl.FC1CNC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)