accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CoH3COOH3COO

CAS RN 61789-51-3

IUPAC Name: λ²-cobalt(2+) bis(3-(3-ethylcyclopentyl)propanoate)
Synonyms: cobalt(II) naphthenate
Molecular Weight (g/mol): 397.42
Molecular Formula: C20H34CoO4
InChi Key: XSWKLHINRKWMTD-UHFFFAOYNA-L
SMILES: [Co++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)