accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 618-32-6

IUPAC Name: benzoyl bromide
Synonyms: benzoyl bromide
Molecular Weight (g/mol): 185.02
Molecular Formula: C7H5BrO
InChi Key: AQIHMSVIAGNIDM-UHFFFAOYSA-N
SMILES: BrC(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results