accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOI

CAS RN 618-51-9

IUPAC Name: 3-iodobenzoic acid
Synonyms: 3-iodobenzoic acid
Molecular Weight (g/mol): 248.02
Molecular Formula: C7H5IO2
InChi Key: KVBWBCRPWVKFQT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(I)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)