accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOIOHI

CAS RN 618-76-8

IUPAC Name: 4-hydroxy-3,5-diiodobenzoic acid
Synonyms: 4-hydroxy-3,5-diiodobenzoic acid
Molecular Weight (g/mol): 389.92
Molecular Formula: C7H4I2O3
InChi Key: XREKTVACBXQCSB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=C(O)C(I)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results