accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NOONOO

CAS RN 618-87-1

IUPAC Name: 3,5-dinitroaniline
Synonyms: 3,5-dinitroaniline
Molecular Weight (g/mol): 183.12
Molecular Formula: C6H5N3O4
InChi Key: MPBZUKLDHPOCLS-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results