accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 618-88-2

IUPAC Name: 5-nitrobenzene-1,3-dicarboxylic acid
Synonyms: 5-nitrobenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 211.13
Molecular Formula: C8H5NO6
InChi Key: NBDAHKQJXVLAID-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1)C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results