accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NHONOO

CAS RN 618-94-0

IUPAC Name: 3-nitrobenzohydrazide
Synonyms: 3-nitrobenzoylhydrazine
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: NQEWXLVDAVTOHM-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results