accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 618-95-1

IUPAC Name: methyl 3-nitrobenzoate
Synonyms: methyl 3-nitrobenzoate
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: AXLYJLKKPUICKV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results