accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6184-17-4

IUPAC Name: sodium (2R,3R,4S)-1-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-3,4-dihydroxy-5-(phosphonooxy)pentan-2-olate dihydrate
Synonyms: sodium (2R,3R,4S)-1-{7,8-dimethyl-2,4-dioxo-3H-benzo[g]pteridin-10-yl}-3,4-dihydroxy-5-(phosphonooxy)pentan-2-olate dihydrate
Molecular Weight (g/mol): 514.36
Molecular Formula: C17H24N4NaO11P
InChi Key: DSUQHAJMVCHMFF-VLRKVHMYSA-N
SMILES: O.O.[Na+].CC1=C(C)C=C2N(C[C@@H]([O-])[C@@H](O)[C@@H](O)COP(O)(O)=O)C3=NC(=O)NC(=O)C3=NC2=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)