accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 619-04-5

CH3H3COO

CAS RN 619-04-5

IUPAC Name: 3,4-dimethylbenzoate
Synonyms: 3,4-dimethylbenzoate
Molecular Weight (g/mol): 149.17
Molecular Formula: C9H9O2
InChi Key: OPVAJFQBSDUNQA-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1C)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)