accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClNOO

CAS RN 619-10-3

IUPAC Name: 2-chloro-5-nitrophenol
Synonyms: 2-chloro-5-nitrophenol
Molecular Weight (g/mol): 173.55
Molecular Formula: C6H4ClNO3
InChi Key: BUMGQSCPTLELLS-UHFFFAOYSA-N
SMILES: OC1=CC(=CC=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results