accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 619-14-7

IUPAC Name: 3-hydroxy-4-nitrobenzoic acid
Synonyms: 3-hydroxy-4-nitrobenzoic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: XLDLRRGZWIEEHT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results