accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 619-45-4

CH3OONH2

CAS RN 619-45-4

IUPAC Name: methyl 4-aminobenzoate
Synonyms: methyl 4-aminobenzoate methyl P-aminobenzoate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: LZXXNPOYQCLXRS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)