accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 619-58-9

IUPAC Name: 4-iodobenzoic acid
Synonyms: 4-iodobenzoic acid
Molecular Weight (g/mol): 248.02
Molecular Formula: C7H5IO2
InChi Key: GHICCUXQJBDNRN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(I)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results