accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 619-84-1

IUPAC Name: 4-(dimethylamino)benzoic acid
Synonyms: 4-(dimethylamino)benzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: YDIYEOMDOWUDTJ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results