CAS RN 61909-81-7
CAS RN 61909-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Solutol HS-15 (Technical Grade), TRC
CAS: 61909-81-7 Molecular Formula: (C2H4O)nC18H36O3 Molecular Weight (g/mol): 344.5 Synonym: Polyethylene Glycol 12-hydroxystearate,Kolliphor HS 15,Solutol HS 15 IUPAC Name: 13-hydroxy-1-(2-hydroxyethoxy)nonadecan-2-one SMILES: CCCCCCC(O)CCCCCCCCCCC(=O)COCCO