accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2OHHO

CAS RN 62-31-7

IUPAC Name: hydrogen 4-(2-aminoethyl)benzene-1,2-diol chloride
Synonyms: dopamine hydrochloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C8H12ClNO2
InChi Key: CTENFNNZBMHDDG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)