accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaCaOONNOOOOOO

CAS RN 62-33-9

IUPAC Name: calcium disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: calcium disodium edta
Molecular Weight (g/mol): 374.27
Molecular Formula: C10H12CaN2Na2O8
InChi Key: SHWNNYZBHZIQQV-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results