accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 620-40-6

IUPAC Name: tribenzylamine
Synonyms: tribenzylamine
Molecular Weight (g/mol): 287.41
Molecular Formula: C21H21N
InChi Key: MXHTZQSKTCCMFG-UHFFFAOYSA-N
SMILES: C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results