accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHH3C

CAS RN 620-93-9

IUPAC Name: 4-methyl-N-(4-methylphenyl)aniline
Synonyms: 4-methyl-N-(4-methylphenyl)aniline
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: RHPVVNRNAHRJOQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C(C)C=C2)C=C1
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results